EMAIL THIS PAGE TO A FRIEND
Peptides
Preliminary analgesic properties of deltorphin-5-methoxytryptamine chimeric opioid peptides.
PMID 21310204
Abstract
To further understand the relationship between melatonin (MT) and deltorphins (Dels) in pain modulation, two chimeric peptides (Del I-5-methoxytryptamine and Del II-5-methoxytryptamine) both containing 5-methoxytryptamine at the carboxyl-terminal of Dels mimicking MT were designed, synthesized and characterized by tail-flick assay in mice. Results showed that intracerebroventricular (i.c.v.) administration of Del I-5-methoxytryptamine (YaFDVVG-X, X is 5-methoxytryptamine, 5, 50 nmol/kg) or Del II-5-methoxytryptamine (YaFEVVG-X, X is 5-methoxytryptamine, 5, 50 nmol/kg) produced stronger analgesia than deltorphins (Del I or Del II alone), and acting even longer and stronger than cocktails containing Del I or Del II (50 nmol/kg) and MT (50 nmol/kg). Naloxone (i.p., 100 nmol/kg) could totally block the analgesic effects induced by the chimeric peptides, while luzindole (specific antagonist of melatonin receptor, i.p., 250 nmol/kg) could only partially inhibit the effects down to that induced by Dels alone. Interestingly, Del I-5-methoxytryptamine and Del II-5-methoxytryptamine act weaker with δ receptor than Dels in vitro but could induce much longer analgesia through co-activating δ opioid receptor and melatonin receptor.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
286583
5-Methoxytryptamine, 97%
C11H14N2O