EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical sciences
A novel solubility-enhanced curcumin formulation showing stability and maintenance of anticancer activity.
PMID 21312196
Abstract
Curcumin (CUR) is an active food compound, but its insolubility and instability in water contributes to low bioavailability. In this study, the solubility of CUR was enhanced by utilizing the solubilizing properties of rubusoside (RUB). The solubility of CUR in water increased linearly from 61 μg/mL to 2.318 mg/mL in the presence of RUB ranging from 1% to 10% (w/v). Dynamic light scattering and transmission electron microscopy studies found that CUR and RUB formed CUR-RUB nanoparticle (∼8 nm) complexes. The RUB-solubilized CUR was stable in physiological conditions and did not precipitate when diluted or degrade when spray-dried to a completely reconstitutable powder. Furthermore, cell viability assays demonstrated the efficacy of RUB-solubilized CUR against human colon, breast, and pancreatic cancer cell lines. The development of this new solubilized, stable, and biologically active CUR formulation lays the foundation for future bioavailability improvement.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
62933
Rubusoside, analytical standard
C32H50O13