EMAIL THIS PAGE TO A FRIEND
Cutis
Cyclosporine for SJS/TEN: a case series and review of the literature.
PMID 21323097
Abstract
Clear guidelines for the treatment of Stevens-Johnson syndrome (SJS) and toxic epidermal necrolysis (TEN) are lacking due to its infrequency and the absence of large controlled studies. Systemic corticosteroids and intravenous immunoglobulin (IVIG) have received considerable attention, though reports of the use of these agents have demonstrated mixed success rates in improving morbidity and mortality from SJS/TEN. We present a case series of 4 patients with SJS/TEN who rapidly responded to treatment with cyclosporin A (CsA). We discuss the proposed mechanism of action and the rationale for the use of cyclosporin based on the currently understood pathophysiologic mechanism of TEN.
Related Materials