EMAIL THIS PAGE TO A FRIEND
Phytochemistry
Lycorine induces cell death in the amitochondriate parasite, Trichomonas vaginalis, via an alternative non-apoptotic death pathway.
PMID 21324496
Abstract
In this study, the mechanism of action of the pro-apoptotic alkaloid lycorine on an amitochondriate cell, the parasite Trichomonas vaginalis, was investigated. The cytotoxicity of lycorine against T. vaginalis was studied from 2.5 to 1000μM and several important ultrastructural alterations were observed by electron microscopy. Lycorine arrested the T. vaginalis cell cycle, although no hallmarks of apoptosis, such as apoptotic bodies, were observed. Consequently, the underlying mechanism of action fails to completely fulfill the criteria for apoptosis. However, some similarities to paraptotic cell death were observed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L5139
Lycorine hydrochloride, ≥98% (TLC), powder
C16H17NO4 · HCl