EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
A stereoselective, Sm(II)-mediated approach to decorated cis-hydrindanes: synthetic studies on faurinone and pleuromutilin.
PMID 21327225
Abstract
The cis-hydrindane motif is found in a number of natural products that display important biological activity. A flexible, stereoselective approach to the framework has been developed that features highly diastereoselective, SmI(2)-mediated cyclisations. The strategy has been exploited in the first synthesis of the proposed structure of faurinone and an approach to the skeleton of the antibacterial natural product, pleuromutilin.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0285
Pleuromutilin, ≥95%
C22H34O5