EMAIL THIS PAGE TO A FRIEND
European spine journal : official publication of the European Spine Society, the European Spinal Deformity Society, and the European Section of the Cervical Spine Research Society
Evaluation of tenoxicam on prevention of arachnoiditis in rat laminectomy model.
PMID 21327813
Abstract
Post laminectomy arachnoiditis has been shown by experiments with rats and post operative radiological imaging in humans. The purpose of this experimental study was to determine the efficacy of tenoxicam in preventing arachnoiditis in rats. Twenty-four Wistar rats were divided into two groups, and L3 laminectomy was performed. In the tenoxicam group, 0.5 mg/kg tenoxicam was applied intraperitoneally. Normal saline was applied intraperitoneally in the control group. Later, the rats were killed at weeks 3 and 6, and the laminectomy sites were evaluated pathologically for arachnoiditis. The results showed that 6 weeks after surgery, the tenoxicam group showed lowest arachnoiditis grades. However, statistically significant difference was not found in arachnoiditis between the control group and the tenoxicam group. Based on these findings it is concluded that application of the tenoxicam after lumbar laminectomy did not effectively reduce arachnoiditis. Performing the most effective surgical technique without damage around tissue in a small surgical wound and having meticulous hemostasis in surgery seem to be the key for preventing arachnoiditis effectively.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T0909
Tenoxicam, NSAID
C13H11N3O4S2
T0040800
Tenoxicam, European Pharmacopoeia (EP) Reference Standard
C13H11N3O4S2