EMAIL THIS PAGE TO A FRIEND
Organic letters
Gold-catalyzed synthesis of benzil derivatives and α-keto imides via oxidation of alkynes.
PMID 21332143
Abstract
An efficient process based on the gold-catalyzed redox reaction has been developed to oxidize 1,2-diarylacetylene or ynamide to 1,2-diaryldiketone or α-keto imide respectively. This process can tolerate a variety of functional groups and affords 1,2-dicarbonyl compounds in excellent yields under mild reaction conditions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B5151
Benzil, 98%
C14H10O2
78600
Phenylglyoxal monohydrate, crystallized, ≥97.0% (GC)
C8H6O2 · H2O