EMAIL THIS PAGE TO A FRIEND
Autonomic & autacoid pharmacology
Colon-specific contractile responses to tetrodotoxin in the isolated mouse gastrointestinal tract.
PMID 21332638
Abstract
1 Tetrodotoxin (TTX) is a useful pharmacological tool for distinguishing neural and myogenic responses of isolated visceral organs to drugs. Although TTX does not generally affect smooth muscle tonus, in this study, we have found that TTX causes contraction of the mouse colon. The aim of this study was to characterize this TTX-induced contraction in the mouse gastrointestinal tract. 2 Longitudinal and circular muscle strips from the stomach and small intestine were less sensitive to TTX. However, TTX contracted both smooth muscle strips from the proximal colon and distal colon. 3 Pretreatment with TTX, Nω -nitro-L-arginine methyl ester (L-NAME), 1H-[1,2,4]oxadiazolo[4,3-a]quinoxalin-1-one (ODQ) and apamin inhibited the TTX-induced contraction. L-NAME, ODQ or apamin itself caused contraction in the colon but not in the gastric and small intestinal strips. Region dependency of L-NAME, ODQ and apamin-induced contraction correlated with that of TTX-induced contraction. 4 L-arginine but not D-arginine inhibited contractility of the colonic strips without affecting the contractility of muscle strips from other regions. Sodium nitroprusside caused strong relaxation of the colonic strips. 5 1,1-dimethyl-4-phenylpiperazinium (DMPP) caused relaxation of proximal and distal colons, which was significantly decreased by L-NAME or apamin. 6 In conclusion, among mouse gastrointestinal preparations, TTX induces contraction of colonic strips preferentially through blockade of potent tonic inhibitory neural outflow, which involves nitrergic and apamin-sensitive pathways. Colon-specific responses to L-arginine, L-NAME, ODQ and apamin support the hypothesis that there is a continuous suppression of colonic motility by enteric inhibitory neurons.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D5891
1,1-Dimethyl-4-phenylpiperazinium iodide, ≥98% (TLC or titration), powder
C12H19IN2