EMAIL THIS PAGE TO A FRIEND
Surgery
Long-term oxandrolone treatment increases muscle protein net deposition via improving amino acid utilization in pediatric patients 6 months after burn injury.
PMID 21333314
Abstract
We recently showed that mechanisms of protein turnover in skeletal muscle are unresponsive to amino acid (AA) infusion in severely burned pediatric patients at 6 months postinjury. In the current study, we evaluated whether oxandrolone treatment affects mechanisms of protein turnover in skeletal muscle and whole-body protein breakdown in pediatric burn patients 6 months postinjury. At the time of admission, patients were randomized to control or oxandrolone treatments. The treatment regimens were continued until 6 months postinjury, at which time patients (n = 26) underwent study with a stable isotope tracer infusion to measure muscle and whole-body protein turnover. Protein kinetics in leg muscle were expressed in nmol/min per 100 mL leg volume (mean ± SE). During AA infusion, rates of protein synthesis in leg muscle were increased (P < .05) in both groups (basal vs AA: control, 51 ± 8 vs 86 ± 21; oxandrolone, 56 ± 7 vs 96 ± 12). In the control group, there was also a simultaneous increase in breakdown (basal vs AA: 65 ± 10 vs 89 ± 25), which resulted in no change in the net balance of leg muscle protein (basal vs AA: -15 ± 4 vs -2 ± 10). In the oxandrolone group, protein breakdown did not change (basal vs AA: 80 ± 12 vs 77 ± 9), leading to increased net balance (basal vs AA: -24 ± 7 vs 19 ± 7; P < .05). Protein breakdown at the whole-body level was not different between the groups. Long-term oxandrolone treatment increased net deposition of leg muscle protein during AA infusion by attenuating protein breakdown, but did not affect whole-body protein breakdown.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0437
Oxandrolone, ≥98% (HPLC)
C19H30O3