EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Planar triazinium cations from VO(2+)-assisted ring cyclizations: a remarkably efficient thiazole species for nuclear staining, PDT and anaerobic photocleavage of DNA.
PMID 21336383
Abstract
Planar triazinium cationic species, from VO(2+)-assisted cyclization of 1-(2-thiazolylazo)-2-naphthol, shows efficient DNA intercalative binding, visible light-induced anaerobic plasmid DNA photocleavage activity and photocytotoxicity in HeLa and MCF-7 cancer cells by an apoptotic pathway with selective localization of the compound in the nucleus as evidenced from the nuclear staining and confocal imaging.
Related Materials