EMAIL THIS PAGE TO A FRIEND
Journal of Huazhong University of Science and Technology. Medical sciences = Hua zhong ke ji da xue xue bao. Yi xue Ying De wen ban = Huazhong keji daxue xuebao. Yixue Yingdewen ban
Application of decellularized scaffold combined with loaded nanoparticles for heart valve tissue engineering in vitro.
PMID 21336730
Abstract
The purpose of this study was to fabricate decelluarized valve scaffold modified with polyethylene glycol nanoparticles loaded with transforming growth factor-β1 (TGF-β1), by which to improve the extracellular matrix microenvironment for heart valve tissue engineering in vitro. Polyethylene glycol nanoparticles were obtained by an emulsion-crosslinking method, and their morphology was observed under a scanning electron microscope. Decelluarized valve scaffolds, prepared by using trypsinase and TritonX-100, were modified with nanoparticles by carbodiimide, and then TGF-β1 was loaded into them by adsorption. The TGF-β1 delivery of the fabricated scaffold was measured by asing enzyme-linked immunosorbent assay. Whether unseeded or reseeded with myofibroblast from rats, the morphologic, biochemical and biomechanical characteristics of hybrid scaffolds were tested and compared with decelluarized scaffolds under the same conditions. The enzyme-linked immunosorbent assay revealed a typical delivery of nanoparticles. The morphologic observations and biological data analysis indicated that fabricated scaffolds possessed advantageous biocompatibility and biomechanical property beyond decelluarized scaffolds. Altogether this study proved that it was feasible to fabricate the hybrid scaffold and effective to improve extracellular matrix microenvironment, which is beneficial for an application in heart valve tissue engineering.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C106402
N-Cyclohexyl-N′-(2-morpholinoethyl)carbodiimide methyl-p-toluenesulfonate, 95%
C14H26N3O · C7H7O3S