EMAIL THIS PAGE TO A FRIEND
Molecular Reproduction and Development
The role of HIRA and maternal histones in sperm nucleus decondensation in the gibel carp and color crucian carp.
PMID 21337451
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
06-598
Anti-acetyl-Histone H4 Antibody, clone, 1 mg/mL, Upstate®