EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Synthesis of 3-indole derivatives by copper sulfonato Salen catalyzed three-component reactions in water.
PMID 21340052
Abstract
An efficient three-component reaction of indole, aldehyde, and malononitrile in water catalyzed by a copper(II) sulfonato Salen complex afforded 3-indole derivatives in good to excellent yields up to 97%.
Related Materials