EMAIL THIS PAGE TO A FRIEND
Organic letters
Synthetic studies on indolocarbazoles: total synthesis of staurosporine aglycon.
PMID 21341756
Abstract
A synthesis of staurosporine aglycon and its analogs was achieved in a 28-36% overall yield starting from 2-methylindole. The prominent key steps for the synthesis of the indolocarbazole alkaloids involved electrocyclization and nitrene insertion reactions.