EMAIL THIS PAGE TO A FRIEND
Organic letters
Charge mapping in 3(10)-helical peptide chains by oxidation of the terminal ferrocenyl group.
PMID 21341757
Abstract
Two series of 3(10)-helical peptides of different lengths and rigidity, based on the strongly foldameric α-aminoisobutyric acid and containing a terminal ferrocenyl unit, have been synthesized. Oxidation-state sensitive spectroscopic tags of helical peptides, the N-H groups, allowed mapping of the charge delocalization triggered by oxidation of the terminal ferrocenyl moiety and were monitored by IR spectroelectrochemistry.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
850993
2-Aminoisobutyric acid, 98%
C4H9NO2