EMAIL THIS PAGE TO A FRIEND
The Journal of dermatology
Angiolymphoid hyperplasia with eosinophilia on the leg successfully treated with T-helper cell 2 cytokine inhibitor suplatast tosilate.
PMID 21342240