EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Highly diastereoselective C(sp3)-C(sp) cross-coupling reactions between 1,3- and 1,4-substituted cyclohexylzinc reagents and bromoalkynes through remote stereocontrol.
PMID 21344579
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N1501
Neocuproine, ≥98%
C14H12N2