EMAIL THIS PAGE TO A FRIEND
Journal of separation science
Simultaneous analysis of hydrochlorothiazide, triamterene and reserpine in rat plasma by high performance liquid chromatography and tandem solid-phase extraction.
PMID 21344645
Abstract
A tandem solid-phase extraction method (SPE) of connecting two different cartridges (C(18) and MCX) in series was developed as the extraction procedure in this article, which provided better extraction yields (>86%) for all analytes and more appropriate sample purification from endogenous interference materials compared with a single cartridge. Analyte separation was achieved on a C(18) reversed-phase column at the wavelength of 265 nm by high-performance liquid chromatography (HPLC). The method was validated in terms of extraction yield, precision and accuracy. These assays gave mean accuracy values higher than 89% with RSD values that were always less than 3.8%. The method has been successfully applied to plasma samples from rats after oral administration of target compounds.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T4143
Triamterene, ≥99%
C12H11N7