EMAIL THIS PAGE TO A FRIEND
Chemical research in toxicology
One-electron reduction of N-chlorinated and N-brominated species is a source of radicals and bromine atom formation.
PMID 21344936
Abstract
Hypochlorous (HOCl) and hypobromous (HOBr) acids are strong bactericidal oxidants that are generated by the human immune system but are implicated in the development of many human inflammatory diseases (e.g., atherosclerosis, asthma). These oxidants react readily with sulfur- and nitrogen-containing nucleophiles, with the latter generating N-halogenated species (e.g., chloramines/bromamines (RR'NX; X = Cl, Br)) as initial products. Redox-active metal ions and superoxide radicals (O(2)(•-)) can reduce N-halogenated species to nitrogen- and carbon-centered radicals. N-Halogenated species and O(2)(•-) are generated simultaneously at sites of inflammation, but the significance of their interactions remains unclear. In the present study, rate constants for the reduction of N-halogenated amines, amides, and imides to model potential biological substrates have been determined. Hydrated electrons reduce these species with k(2) > 10(9) M(-1) s(-1), whereas O(2)(•-) reduced only N-halogenated imides with complex kinetics indicative of chain reactions. For N-bromoimides, heterolytic cleavage of the N-Br bond yielded bromine atoms (Br(•)), whereas for other substrates, N-centered radicals and Cl(-)/Br(-) were produced. High-level quantum chemical procedures have been used to calculate gas-phase electron affinities and aqueous solution reduction potentials. The effects of substituents on the electron affinities of aminyl, amidyl, and imidyl radicals are rationalized on the basis of differential effects on the stabilities of the radicals and anions. The calculated reduction potentials are consistent with the experimental observations, with Br(•) production predicted for N-bromosuccinimide, while halide ion formation is predicted in all other cases. These data suggest that interaction of N-halogenated species with O(2)(•-) may produce deleterious N-centered radicals and Br(•).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
178098
Glutarimide, 98%
C5H7NO2