EMAIL THIS PAGE TO A FRIEND
European journal of medicinal chemistry
2-Acylamino-5-chlorobenzophenones with enhanced selectivity towards malaria parasites.
PMID 21345543
Abstract
Previously we described a series of 5-acylaminobenzophenones with considerable antimalarial activity. Unfortunately, most compounds also displayed high cytotoxicity resulting in low selectivity towards malaria parasites. Through the replacement of the 5-acylamino moiety by simple chlorine and further modifications of the 2-acylamino residue we could obtain inhibitors with improved selectivity towards malaria parasites combined with an acceptable reduction of antimalarial activity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A45564
2-Amino-5-chlorobenzophenone, 98%
C13H10ClNO