EMAIL THIS PAGE TO A FRIEND
Nihon Hotetsu Shika Gakkai zasshi
[A light-cured acrylic adhesive for fixing resin retention devices to the wax pattern].
PMID 2134790
Abstract
A light-cured acrylic adhesive for fixing resin retention devices to the wax pattern was prepared. The adhesive consisted of trimethylolpropane triacrylate, 2-ethylhexyl acrylate, benzoin methyl ether, p dimethylaminobenzaldehyde and p-methoxyphenol. The adhesive could be cured within 20 sec not only by an UV photo curing unit but by a visible-light source with a xenon lamp. The adhesive and retention beads burned out after about an hour in the electric furnace at 400 c. The metal specimens with retention devices were cast in Ag-Pd-Cu-Au alloy with the use of two types of retention beads adhesive. The light-cured adhesive was superior to the conventional one in handling and some other properties. This adhesive may be used to fabricate composite veneered prostheses with minimum errors in laboratory procedure.