EMAIL THIS PAGE TO A FRIEND
Yao xue xue bao = Acta pharmaceutica Sinica
Development and validation of HPLC-UV-MS method for the control of four anti-diabetic drugs in suspected counterfeit products.
PMID 21348424
Abstract
An HPLC-UV method has been developed for the determination of valibose, miglitol, voglibose and acarbose, the four anti-diabetic drugs. The separation was accomplished successfully by using reversed phase chromatography (Prevail carbohydrate column, 250 mm x 4.6 mm, 5 microm) with a gradient acetonitrile-phosphate buffer solution (pH 8.0) at a wavelength of 210 nm. Furthermore, the method of a high-performance liquid chromatography coupled with ESI-MS in positive ionization mode has been established. These two methods were successfully applied to the assay and qualitative detection of four alpha-glucosidase inhibitors in the potential counterfeit anti-diabetic drugs.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M1574 Miglitol
C8H17NO5
50359
Voglibose, ≥97.0% (TLC)
C10H21NO7