EMAIL THIS PAGE TO A FRIEND
Reproductive biology and endocrinology : RB&E
Characterization of bovine immortalized luteal endothelial cells: action of cytokines on production and content of arachidonic acid metabolites.
PMID 21349168
Abstract
The interactions between luteal, vascular endothelial, immune cells and its products: steroids, peptide hormones, prostaglandins (PGs), growth factors and cytokines play a pivotal role in the regulation of corpus luteum (CL) function. Luteal endothelial cells undergo many dynamic morphological changes and their action is regulated by cytokines. The aims are: (1) to establish in vitro model for bovine luteal endothelial cells examination; (2) to study the effect of cytokines: tumor necrosis factor alpha (TNFalpha) and interferon gamma (IFNgamma) on cell viability, leukotrienes (LTs) and PG synthases, and endothelin-1 (EDN-1) mRNA, protein expression and their secretion in bovine immortalized luteal endothelial (EnCL-1) cells. The primary cultures of bovine luteal endothelial cells were immortalized by transfection with vector carrying the Simian virus 40 T-antigen (SV40 T-ag) sequence. Expression of SV40 T-ag gene in EnCL-1 cells was confirmed by RT-PCR and immunofluorescence staining showed the presence of endothelial cell markers: VE-cadherin and von Willebrand factor. EnCL-1 cells were stimulated by TNFalpha with IFNgamma (50 ng/ml each) for 24 h. Cell viability, mRNA expression (real time RT-PCR), protein expression (western blotting) for LTC4 synthase (LTC4S), LTA4 hydrolase (LTA4H), PGE2 and PGF2alpha synthases and endothelin-1 (EDN-1), and levels of LTs (B4 and C4) and PGs (E2 and F2alpha) and EDN-1 in the medium (EIA) were evaluated. We received immortalized luteal endothelial cell line (EnCL-1). Cytokines did not change EnCL-1 cell viability but increased mRNA expression of LTC4S, LTA4H, PGE2 and PGF2alpha synthases and EDN-1. EDN-1/2/3, LTC4 and PGF2alpha synthases protein expression were elevated in the presence of TNFalpha/IFNgamma, and accompanied by increased EDN-1, LTC4 and PGF2alpha secretion. Cytokines had no effect on PGES and LTA4H protein expression, and PGE2 and LTB4 release. TNFalpha and IFNgamma modulate EnCL-1 cell function. Moreover, established EnCL-1 cell line appears to be a good model for investigating the molecular mechanisms related to cytokines action and aa metabolites production in cattle.