EMAIL THIS PAGE TO A FRIEND
QJM : monthly journal of the Association of Physicians
Lercanidipine-associated nocturia.
PMID 21349908
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L6668
Lercanidipine hydrochloride, ≥98% (HPLC)
C36H41N3O6 · HCl