EMAIL THIS PAGE TO A FRIEND
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica
[Effect of three penetration enhancers on corneal permeability of mangiferin in vitro].
PMID 21355232
Abstract
To investigate the effects of labrasol, solutol HS 15 and transcutol P on the corneal permeability of mangiferin in vitro. The effects of three penetration enhancers on the corneal permeability of mangiferin were investigated in vitro by using isolated rabbit corneas. The apparent Papp enhancements were increased 1.80, 3.27, 3.41 and 4.76-folds with Lab at 1.0%, 1.5%, 2.0% and 3.0% (P < 0.01), respectively. The apparent Papp increased 1.98 and 3.07-folds with Sol at 0.2% and 0.4% (P < 0.01), respectively, but reduced with 0.010%-0.03% Trans. The Papp value of mangiferin is significantly enhanced by 1.0%-3.0% Lab, 0.2% and 0.4% Sol, however the Papp value of mangiferin is reduced by 0.01%-0.03% Trans.
Related Materials