EMAIL THIS PAGE TO A FRIEND
International journal of urology : official journal of the Japanese Urological Association
Editorial comment from Dr Dmochowski to propiverine hydrochloride in Japanese patients with overactive bladder: a randomized, double-blind, placebo-controlled trial.
PMID 21355895
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0602 Propiverine hydrochloride, ≥98% (HPLC)
C23H29NO3 · HCl