EMAIL THIS PAGE TO A FRIEND
Cell metabolism
Regulation of lipid homeostasis by the bifunctional SREBF2-miR33a locus.
PMID 21356514
Abstract
The sterol regulatory element-binding factor-2 (SREBF2) gene is a bifunctional locus encoding SREBP-2, a well-known transcriptional regulator of genes involved in cholesterol biosynthesis, and microRNA-33a, which has recently been shown to reduce expression of proteins involved in export of cholesterol and β-oxidation of fatty acids, thus adding an unexpected layer of complexity and fine-tuning to regulation of lipid homeostasis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AV09037
Anti-SREBF2 antibody produced in rabbit, affinity isolated antibody