PloS one
The human serum metabolome.
PMID 21359215
Abstract
Continuing improvements in analytical technology along with an increased interest in performing comprehensive, quantitative metabolic profiling, is leading to increased interest pressures within the metabolomics community to develop centralized metabolite reference resources for certain clinically important biofluids, such as cerebrospinal fluid, urine and blood. As part of an ongoing effort to systematically characterize the human metabolome through the Human Metabolome Project, we have undertaken the task of characterizing the human serum metabolome. In doing so, we have combined targeted and non-targeted NMR, GC-MS and LC-MS methods with computer-aided literature mining to identify and quantify a comprehensive, if not absolutely complete, set of metabolites commonly detected and quantified (with today's technology) in the human serum metabolome. Our use of multiple metabolomics platforms and technologies allowed us to substantially enhance the level of metabolome coverage while critically assessing the relative strengths and weaknesses of these platforms or technologies. Tables containing the complete set of 4229 confirmed and highly probable human serum compounds, their concentrations, related literature references and links to their known disease associations are freely available at http://www.serummetabolome.ca.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H9521
15(S)-Hydroperoxy-(5Z,8Z,11Z,13E)-eicosatetraenoic acid, ~100 μg/mL in ethanol, ≥95%
C20H32O4
H1142
15(S)-Hydroxy-(5Z,8Z,11Z,13E)-eicosatetraenoic acid, ~150 μg/mL in ethanol, ≥97.0% (HPLC)
C20H32O3
D8440
15-Deoxy-Δ12,14-prostaglandin J2, ≥95% (HPLC), 1 mg/mL in methyl acetate
C20H28O3
A8973
2-Arachidonyl glycerol, ~10 mg/mL, ≥98% (HPLC)
C23H38O4
I9030
2-Propanol, meets USP testing specifications
C3H8O
650447
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
C3H8O
34863
2-Propanol, for HPLC, 99.9%
C3H8O
439207
2-Propanol, for HPLC, 99.5%
C3H8O
190764
2-Propanol, ACS reagent, ≥99.5%
C3H8O
673773
2-Propanol, ACS reagent, ≥99.5%
C3H8O
59300
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)
C3H8O
33539
2-Propanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
C3H8O
24137
2-Propanol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC)
C3H8O
109827
2-Propanol, Laboratory Reagent, ≥99.5%
C3H8O
C6046
20-Carboxy-leukotriene B4, ~1 mg/mL in ethanol, ≥90% (HPLC)
C20H30O6
H3023
20-Hydroxy-(5Z,8Z,11Z,14Z)-eicosatetraenoic acid, ~100 μg/mL in ethanol, ≥90% (HPLC)
C20H32O3
166898
3-Hydroxybutyric acid, 95%
C4H8O3
H4019
8(S)-Hydroxy-(5Z,9E,11Z,14Z)-eicosatetraenoic acid, ~100 μg/mL in ethanol, ~98% (HPLC)
C20H32O3
07691
9-cis,11-trans-Octadecadienoic acid solution
C18H32O2
323594
α-Hydroxyisobutyric acid, 99%
C4H8O3
164976
α-Hydroxyisobutyric acid, 98%
C4H8O3
695092
Acetic acid, ACS reagent, ≥99.7%
C2H4O2
A6283
Acetic acid, ReagentPlus®, ≥99%
C2H4O2
338826
Acetic acid, ≥99.99% trace metals basis
C2H4O2
33209
Acetic acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
C2H4O2
27225
Acetic acid, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
C2H4O2
27221
Acetic acid, puriss., 99-100%
C2H4O2
45754
Acetic acid solution, for HPLC
C2H4O2
650501
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
C3H6O
270725
Acetone, for HPLC, ≥99.9%
C3H6O