EMAIL THIS PAGE TO A FRIEND
Dalton transactions (Cambridge, England : 2003)
A soluble form of nano-sized colloidal manganese(IV) oxide as an efficient catalyst for water oxidation.
PMID 21359392
Abstract
A soluble form of colloidal manganese(IV) oxide showed efficient oxygen evolution or water oxidation in presence of oxone, H(2)O(2), cerium(IV) ammonium nitrate and tris(2,2'-bipyridyl)ruthenium(III).
Related Materials