EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
Stereoselective rearrangement of guaianolides to tricyclic δ-valerolactones.
PMID 21359403
Abstract
An unprecedented, highly stereoselective rearrangement of guaianolides, bearing a double bond at the C-6/C-6a position, to tricyclic δ-valerolactones is described.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
389579
δ-Valerolactone, technical grade
C5H8O2