EMAIL THIS PAGE TO A FRIEND
Cellular and molecular neurobiology
Time course of postnatal distribution of doublecortin immunoreactive developing/maturing neurons in the somatosensory cortex and hippocampal CA1 region of C57BL/6 mice.
PMID 21360195
Abstract
In this study, we observed neuroblast differentiation in the somatosensory cortex (SSC) and hippocampal CA1 region (CA1), which is vulnerable to oxidative stress, of the mouse at various early postnatal days (P) 1, 7, 14, and 21 using doublecortin (DCX, a marker for neuroblasts). Cresyl violet and NeuN (Neuronal Nuclei) staining showed development of layers as well as neurons in the SSC and CA1. At P1, DCX-positive neuroblasts expressed strong DCX immunoreactivity in both the SSC and CA1. Thereafter, DCX immunoreactivity was decreased with time. At P7, many DCX-immunoreactive neuroblasts were well detected in the SSC and CA1. At P14, some DCX-positive neuroblasts were found in the SSC and CA1: The immunoreactivity was weak. At P21, DCX immunoreactivity was hardly found in cells in the SSC and CA1. These results suggest that DCX-positive neuroblasts were significantly decreased in the mouse SSC and CA1 from P14.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C5042
Cresyl Violet acetate, certified by the Biological Stain Commission
C18H15N3O3
255246
Cresyl Violet perchlorate
C16H11N3O · HClO4