EMAIL THIS PAGE TO A FRIEND
Archives of Iranian medicine
Hemiparkinsonism-hemiatrophy syndrome.
PMID 21361726
Abstract
The syndrome of hemiparkinsonism-hemiatrophy is an uncommon form of secondary Parkinsonism that presents with unilateral body Parkinsonism plus variable atrophy on the same side. Diagnosis of this syndrome needs a complete past medical history taking, as well as assessment of the familial history, clinical examination and complete paraclinical tests.The response to medical therapy has been variable in various researches. This case showed a good response to the addition of a dopamine agonist to levodopa therapy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B5311
Biperiden hydrochloride, ≥98% (HPLC), powder
C21H30ClNO