EMAIL THIS PAGE TO A FRIEND
The Veterinary record
Illegal use of Amprol-Plus for game.
PMID 2136331
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
33996
Ethopabate, VETRANAL, analytical standard
C12H15NO4