EMAIL THIS PAGE TO A FRIEND
Natural product communications
Chemical composition of the essential oil of Croton gossypiifolius from Venezuela.
PMID 21366055
Abstract
The essential oil from leaves of Croton gossypiifolius Vahl. (Euphorbiaceae) was obtained by hydrodistillation, and analyzed by GC/FID and GC/MS. The constituents were identified by their mass spectra and Kovats' indices. Fifty-one compounds accounting for 92% of the oil were detected, and 47 of them were identified. The oil was dominated by oxygenated sesquiterpenes with the major presence of alpha-cedrene oxide (18.6%), spathulenol (16.3%), valencene (5.8%), geranyl-pentanoate (5.3%), alpha-cadinol (4.0%), germacrene D (3.5%) and longifolene (3.3%).