EMAIL THIS PAGE TO A FRIEND
American Journal of Physiology: Renal Physiology
Glycated and carbamylated albumin are more "nephrotoxic" than unmodified albumin in the amphibian kidney.
PMID 21367923
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
06-127
Anti-PDGF Antibody, neutralizing, clone, Upstate®, from goat