EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Engineering multifunctional magnetic-quantum dot barcodes by flow focusing.
PMID 21369572
Abstract
A simple one-step flow focusing method was used to embed both magnetic nanoparticles and quantum dots in microbeads in controlled ratios to generate a large library of molecular barcodes for biological applications.