EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
From difluoromethyl 2-pyridyl sulfone to difluorinated sulfonates: a protocol for nucleophilic difluoro(sulfonato)methylation.
PMID 21370337
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
746061
Difluoromethyl 2-pyridyl sulfone, 97% (HPLC)
C6H5F2NO2S