EMAIL THIS PAGE TO A FRIEND
The journal of medical investigation : JMI
Protective effect of photodegradation product of nifedipine against tumor necrosis factor alpha-induced oxidative stress in human glomerular endothelial cells.
PMID 21372496
Abstract
Recently, increasing evidence suggests that the antihypertensive drug nifedipine acts as a protective agent for endothelial cells, and that the activity is unrelated to its calcium channel blocking. Nitrosonifedipine (NO-NIF) is metabolically and photochemically produced from nifedipine, and NO-NIF has been recognized as a contaminant of nifedipine because it has no antihypertensive effect. Treatment of tumor necrosis factor-α (TNF-α) suppressed the cell viability and facilitated the expression of Inter-Cellular Adhesion Molecule 1(ICAM-1) in human glomerular endothelial cells (HGECs) though, pretreatment of NO-NIF significantly recovered the TNF-α-induced cell damage to the same extent as Trolox-C did, and suppressed the ICAM-1 expression in a concentration dependent manner. In addition, NO-NIF inhibited the cell toxicity induced by cumene hydroperoxide, which hampers the integrity of cell membrane through oxidative stress, as effective as Trolox-c. These data suggest that NO-NIF is a candidate for a new class of antioxidative drug that protect cells against oxidative stress in glomerular endothelial cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
247502
Cumene hydroperoxide, technical grade, 80%
C9H12O2