EMAIL THIS PAGE TO A FRIEND
Sheng wu yi xue gong cheng xue za zhi = Journal of biomedical engineering = Shengwu yixue gongchengxue zazhi
[Biological activity evaluation of porous HA ceramics using NH4 HCO3/PVA as pore-creating agents].
PMID 21374979
Abstract
Porous HA ceramics were prepared by using NH4 HCO3/PVA as pore-formed material along with biological glass as intensifier, and these ceramics were immersed in Locke's Physiological Saline and Simulate Body Fluid (SBF). The changes of phase composition, grain size and crystallinity of porous HA ceramics before and after immersion were investigated by X-Ray Diffraction (XRD) and Scanning Electron Microscopy (SEM). The biological activity was evaluated. The porous HA ceramics showed various degrees of decomposition after immersion in the two solution systems, but there was no evident change in respect to crystallinity. Besides, the impact of different degrees of solution systems on the change of grain size and planar preferred orientation was observed. The TCP phase of the ceramics immersed in Locke's Physiological Saline decomposed and there was no crystal growth on the surface of ceramics; however, the grain size of ceramics immersed in SBF became refined in certain degree and the surface of ceramics took on the new crystal growth.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A6141
Ammonium bicarbonate, ReagentPlus®, ≥99.0%
CH5NO3