EMAIL THIS PAGE TO A FRIEND
Journal of pharmacological and toxicological methods
Functional biomarkers of musculoskeletal syndrome (MSS) for early in vivo screening of selective MMP-13 inhibitors.
PMID 21376127
Abstract
Long-term administration of non-selective matrix metalloproteinase (MMP) inhibitors, such as marimastat, in humans elicits musculoskeletal syndrome (MSS), a syndrome characterized by joint damage including pain, stiffness, and inflammation. This pathology is a significant obstacle to the clinical development of MMP inhibitors and in pre-clinical models MSS can be verified only after terminal histopathology. Consequently, we devised a longitudinal and functional readout of MSS in conscious rats treated with marimastat that was validated against terminal histological assessment. MSS was induced by minipump infusion of marimastat (5-10mg/kg/day). In marimastat-treated or vehicle-control groups, three possible functional biomarkers were assessed: paw volume (PV), landing foot splay separation (LFSS), and rotarod performance (n=6 rats/group for each endpoint). Histologically, fibrosis scores in the synovium and ligament increased from 0 on Day 1 (D1) to 4.6±0.2 and 4.7±0.1, respectively, on D15; growth plate thickness was also elevated from 215.0±6.3μm (D1) to 253.3±8.0μm (D15). While neither PV nor LFSS were correlative with MSS histopathology, marimastat (10mg/kg/day) reduced rotarod performance from 180±0s (D0) to 135±30s (D9) using a constant speed protocol (10rpm, 180s) and from 180±0s (D0) to 96±6s (D6) employing a variable speed protocol (increasing from 5 to 25rpm over 180s). Results of the present study demonstrate that rotarod performance can be used as a predictive longitudinal, in vivo functional biomarker of MSS concomitant with histological evidence of joint damage to effectively facilitate compound selection during drug discovery. Moreover, for targets with a mechanistic risk for MSS, the model is also conducive to inclusion in secondary pharmacodynamic studies during lead optimization to identify the best (safest) compounds for advancement into clinical trials.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M2699
Marimastat, ≥98% (HPLC)
C15H29N3O5