Description
Molecular Formula
Add to Cart
36232 Density Standard 1251 kg/m3, H&D Fitzgerald Ltd. Quality
109185
Saccharin, ≥98%
C7H5NO3S
S6014
Sodium bicarbonate, ACS reagent, ≥99.7%
CHNaO3
S8875
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
CHNaO3
900506
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
CHNaO3
36486
Sodium bicarbonate concentrate, 0.1 M NaHCO3 in water, eluent concentrate for IC
243973
Sodium bisulfite, ACS reagent
NaHSO3
799394
Sodium bisulfite, anhydrous, free-flowing, Redi-Dri, ACS reagent
NaHSO3
13438
Sodium bisulfite solution, purum, ~40%
HNaO3S
S4547
Sodium bromide, BioXtra, ≥99.0%
BrNa
71329
Sodium bromide, BioUltra, ≥99.5% (AT)
BrNa
460591
Sodium bromide, anhydrous, beads, −10 mesh, 99.999% trace metals basis
BrNa
229881
Sodium bromide, ≥99.99% trace metals basis
BrNa
310506
Sodium bromide, ACS reagent, ≥99.0%
BrNa
220345
Sodium bromide, ReagentPlus®, ≥99%
BrNa
223484
Sodium carbonate, ACS reagent (primary standard), anhydrous, 99.95-100.05% dry basis
CNa2O3
222321
Sodium carbonate, ACS reagent, anhydrous, ≥99.5%, powder or granules
CNa2O3
223530
Sodium carbonate, powder, ≥99.5%, ACS reagent
CNa2O3
S2127
Sodium carbonate, ReagentPlus®, ≥99.5%
CNa2O3
13418
Sodium carbonate, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, E500, anhydrous, 99.5-100.5% (calc. to the dried substance)
CNa2O3
900503
Sodium carbonate, anhydrous, powder or granules, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
CNa2O3
900505
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
CNa2O3
900504
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent (primary standard), 99.95-100.05% dry basis
CNa2O3
07982
Sodium chloride physiological solution, BioUltra, tablet
52455
Sodium chloride physiological solution, BioUltra, tablet
201154
Sodium fluoride, ACS reagent, ≥99%
FNa
S1504
Sodium fluoride, ReagentPlus®, ≥99%
FNa
30105
Sodium fluoride, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99%
FNa
01148
Sodium fluoride, puriss., meets analytical specification of Ph. Eur., BP, USP, 98.5-100.5% (calc. to the dried substance)
FNa
71710
Sodium iodide, puriss. p.a., ≥99.0% (AT)
INa