EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Lewis acid double metal cyanide catalysts for hydroamination of phenylacetylene.
PMID 21380422
Abstract
Double metal cyanides (DMCs) are highly active recyclable heterogeneous catalysts for hydroamination of phenylacetylene with 4-isopropylaniline. The best hydroamination yields are obtained with Zn-Co DMCs, especially if the particle size is decreased by a reverse emulsion synthesis technique.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
175439
4-Isopropylaniline, 99%
C9H13N