EMAIL THIS PAGE TO A FRIEND
Mikrobiolohichnyi zhurnal (Kiev, Ukraine : 1993)
[Screening of yeast-producers of alpha-L-rhamnosidase].
PMID 21381311
Abstract
Screening of producers of alpha-L-rhamnosidase among 331 yeast strains (representatives of 12 genera and 17 species) has been performed. A capacity to synthesize the enzyme was revealed in 45% of the studied strains with activity 0.01-0.6 un/ml. Complex enzymatic preparations of alpha-L-rhamnosidase were obtained from cultural liquid of two the most active producers by fractionation with ammonium sulfate (90% saturation). The influence of pH and temperature on enzymes activities and also specificity towards natural and synthetic substrates were studied.