EMAIL THIS PAGE TO A FRIEND
Se pu = Chinese journal of chromatography
[Simultaneous determination of delta-9-tetrahydrocannabinol cannabidiol and cannabinol in edible oil using ultra performance liquid chromatography-tandem mass spectrometry].
PMID 21381415
Abstract
A method for the simultaneous determination of delta-9-tetrahydrocannabinol (THC), cannabidiol (CBD) and cannabinol (CBN) in edible oil was developed using ultra performance liquid chromatography-tandem mass spectrometry (UPLC-MS/MS). The target compounds were extracted with methanol, purified by an LC-Alumina-N solid phase extraction cartridge, separated and detected by the UPLC-MS/MS. Quantitative analysis was corrected by an isotope internal standard method using delta-9-THC-D3 as internal standard. Average recoveries for the target compounds varied from 68.0% to 101.6% with the relative standard deviations ranging from 7.0% to 20.1% at three spiked levels. The limits of detection (LOD) of the method were from 0.06-0.17 microg/kg and the limits of quantification (LOQ) were in the range of 0.20-0.52 microg/kg. The results showed that the method is able to meet the requirements for the simultaneous determination of THC, CBD and CBN in edible oil.