EMAIL THIS PAGE TO A FRIEND
Se pu = Chinese journal of chromatography
[Determination of the migration of bisphenol diglycidyl ethers from food contact materials by high performance chromatography-tandem mass spectrometry coupled with multi-walled carbon nanotubes solid phase extraction].
PMID 21381429
Abstract
A comprehensive analytical method based on high performance liquid chromatography-tandem mass spectrometry (HPLC-MS/MS) was developed for measuring 6 exogenous endocrine disruptors--bisphenol diglycidyl ethers, including bisphenol A diglycidyl ether (BADGE), bisphenol A glycidyl (2,3-dihydroxypropyl) ether (BADGE x H2O), bisphenol A glycidyl (3-chloro-2-hydroxypropyl) ether ( BADGE x HCl), bisphenol A (3-chloro-2-hydroxypropyl) (2,3-dihydroxypropyl) ether (BADGE x H2O x HCl), bisphenol F diglycidyl ether (BFDGE) and bisphenol F bis (3-chloro-2-hydroxypropyl) ether (BFDGE x 2HCl). The samples were extracted with methyl tert-butyl ether (MTBE) by ultrasonic wave assistant extraction. The extracts were cleaned up and concentrated on multi-walled carbon nanotubes (MWCNTs). The target compounds were analyzed by HPLC-MS/MS under positive ion mode using a COSMOSIL C18 column as analytical column. Under the optimal conditions, the calibration curves showed a good linearity in the concentration range of 1.0-100.0 microg/L for 6 target compounds. The correlation coefficients (r2) were higher than 0.999 1. Recoveries of 6 analytes at three spiked levels ranged from 78.6% to 89.9%, with relative standard deviations (RSDs) less than 10%. The detection limits of the method ranged from 0.5 to 1.5 microg/L. The method is sensitive and simple, and is suitable for the rapid determination of the migration of bisphenol diglycidyl ethers from food contact materials.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D3415
Bisphenol A diglycidyl ether
C21H24O4
15138
Bisphenol A diglycidyl ether, analytical standard
C21H24O4