EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Synthesis of (-)-homogalanthamine from naltrexone.
PMID 21381712
Abstract
Acetylcholinesterase inhibitor (-)-homogalanthamine 3 was synthesized from μ opioid antagonist naltrexone (2) in 16% total yield. The synthesis features Grob fragmentation as a key reaction, which was especially accelerated in the presence of 15-crown-5.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
188832
15-Crown-5, 98%
C10H20O5