EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Dihydropyranone formation by ipso C-H activation in a glucal 3-carbamate-derived rhodium acyl nitrenoid.
PMID 21381715
Abstract
By using (N-tosyloxy)-3-O-carbamoyl-D-glucal 10, which removes the need for a hypervalent iodine(III) oxidant, we provide evidence for rhodium nitrenoid-mediated ipso C-H activation as the origin of a C3-oxidized dihydropyranone product 3. This system may be especially susceptible to such a pathway because of the ease of forming a cation upon hydride transfer to the rhodium-complexed acyl nitrene.