EMAIL THIS PAGE TO A FRIEND
Journal of clinical virology : the official publication of the Pan American Society for Clinical Virology
Co-infection of arboviruses presenting as Acute Encephalitis Syndrome.
PMID 21382746