EMAIL THIS PAGE TO A FRIEND
Environmental toxicology
Multiple biomarkers responses in juvenile rainbow trout, Oncorhynchus mykiss, after acute exposure to a fungicide propiconazole.
PMID 21384499
Abstract
In this study, the toxic effects of propiconazole (PCZ), a triazole fungicide present in aquatic environment, were studied in juvenile rainbow trout, Oncorhynchus mykiss, by acute toxicity test with the concentration of 5.04 mg/L (96 h LC50). Morphological indices, hematological parameters, liver xenobiotic-metabolizing response, and tissue antioxidant status were evaluated. Compared with the control group, fish exposed to PCZ showed significantly higher Leuko, PCV, MCHC, and hepatic EROD, and significantly lower MCV. CF and HSI were not significantly different among groups. SOD, CAT, GPx, and GR activities increased significantly in liver of experimental groups, but decreased significantly in gill. In general, antioxidant enzyme activity in intestine was less evident than in liver. Oxidative stress indices (levels of LPO and CP) were significantly higher in gill. Additionally, through chemometrics of all parameters measured in this study, two groups with 67.29% of total accumulated variance were distinguished. In short, the physiological and biochemical responses in different tissues of fish indicated that PCZ-induced the stressful environmental conditions. But according to PCZ residual status in the natural environment, more long-term experiments at lower concentrations will be necessary in the future. © 2011 Wiley Periodicals, Inc. Environ Toxicol, 2013.