EMAIL THIS PAGE TO A FRIEND
Organic letters
Access to enantiomerically enriched cis-2,3-disubstituted azetidines via diastereoselective hydrozirconation.
PMID 21384824
Abstract
An asymmetric variant of the hydrozirconation reaction has been established starting from Boc-protected chiral allylic amines. The resulting diastereoselectively formed N-functionalized organozirconiums can be considered as promising chirons. In this case, they have been transformed into enantiomerically enriched cis-2,3-disubstituted azetidines through a iodination/cyclization sequence.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
383287
Zirconium(IV) silicate, −325 mesh
O4SiZr